2-Benzoyl-5-fluorobenzoic acid ≥97%

Supplier: Matrix Scientific

082050-1G
102836-422EA 636.92 USD
102836-422
2-Benzoyl-5-fluorobenzoic acid ≥97%
2-Benzoyl-5-fluorobenzoic acid

Hazard: Irritant

Formula: C₁₄H₉FO₃
MDL Number: MFCD09991767
CAS Number: 142153-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR