3-[(5-Bromopyrimidin-2-yl)oxy]benzoic acid ≥97%

Supplier: Matrix Scientific

082039-1G
102836-398EA 806.15 USD
102836-398
3-[(5-Bromopyrimidin-2-yl)oxy]benzoic acid ≥97%
3-[(5-Bromopyrimidin-2-yl)oxy]benzoic acid

Hazard: Irritant

Formula: C₁₁H₇BrN₂O₃
MDL Number: MFCD09864913
CAS Number: 1086379-59-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR