4-(2-Chloropyrimidin-4-yl)benzoic acid ≥97%
Supplier: Matrix Scientific
082037-1G
102836-394EA
806.15
USD
102836-394
4-(2-Chloropyrimidin-4-yl)benzoic acid ≥97%
4-(2-Chloropyrimidin-4-yl)benzoic acid
Hazard: Irritant
Formula:
C₁₁H₇ClN₂O₂ |
MDL Number:
MFCD09861563 CAS Number: 281232-89-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...