4-(6-Chloropyrazin-2-yl)benzoic acid ≥97%

Supplier: Matrix Scientific

082036-1G
102836-392EA 636.92 USD
102836-392
4-(6-Chloropyrazin-2-yl)benzoic acid ≥97%
4-(6-Chloropyrazin-2-yl)benzoic acid

Hazard: Irritant

Formula: C₁₁H₇ClN₂O₂
MDL Number: MFCD09864912
CAS Number: 1086379-53-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR