3-[2-(Tetrahydro-2H-pyran-2-ylethoxy)phenoxy]-4,4,5,5-tetramethyl[1,3,2]dioxaboralane ≥97%

Supplier: Matrix Scientific

081993-1G
102836-284EA 863.08 USD
102836-284
3-[2-(Tetrahydro-2H-pyran-2-ylethoxy)phenoxy]-4,4,5,5-tetramethyl[1,3,2]dioxaboralane ≥97%
3-[2-(Tetrahydro-2H-pyran-2-ylethoxy)phenoxy]-4,4,5,5-tetramethyl[1,3,2]dioxaboralane

Hazard: Irritant

Formula: C₁₉H₂₉BO₅
MDL Number: MFCD14666469
CAS Number: 1334165-03-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR