tert-Butyl-4-{2-[3-(4,4,5,5-tetramethyl[1,3,2]dioxaborolan-2-yl)phenoxy]ethyl}piperidine-1-carboxylate ≥97%

Supplier: Matrix Scientific

081989-1G
102836-272EA 1118.46 USD
102836-272
tert-Butyl-4-{2-[3-(4,4,5,5-tetramethyl[1,3,2]dioxaborolan-2-yl)phenoxy]ethyl}piperidine-1-carboxylate ≥97%
tert-Butyl-4-{2-[3-(4,4,5,5-tetramethyl[1,3,2]dioxaborolan-2-yl)phenoxy]ethyl}piperidine-1-carboxylate

Hazard: Irritant

Formula: C₂₄H₃₈BNO₅
MW: 431.39 g/mol
MDL Number: MFCD12827860
CAS Number: 1310405-34-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR