4-Piperidin-4-ylbutan-1-amine ≥97%

Supplier: Matrix Scientific

081954-1G
102836-190EA 949.23 USD
102836-190
4-Piperidin-4-ylbutan-1-amine ≥97%
4-Piperidin-4-ylbutan-1-amine

Hazard: Irritant

Formula: C₉H₂₀N₂
MDL Number: MFCD08235201
CAS Number: 71745-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR