tert-Butyl (2R)-4,4-diethoxy-2-(aminomethyl)piperidine-1-carboxylate ≥97%

Supplier: Matrix Scientific

081953-1G
102836-188EA 2846.15 USD
102836-188
tert-Butyl (2R)-4,4-diethoxy-2-(aminomethyl)piperidine-1-carboxylate ≥97%
tert-Butyl (2R)-4,4-diethoxy-2-(aminomethyl)piperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₅H₃₀N₂O₄
MW: 302.42 g/mol
MDL Number: MFCD08056296
CAS Number: 1214977-76-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR