N-(1H-Indol-5-ylmethyl)-N-methylamine ≥97%
Supplier: Matrix Scientific
Synonyms:
1-(1H-Indol-5-yl)-N-methylmethanamine
081942-1G
102836-164EA
806.15
USD
102836-164
N-(1H-Indol-5-ylmethyl)-N-methylamine ≥97%
N-(1H-Indol-5-ylmethyl)-N-methylamine
Hazard: Irritant
Formula:
C₁₀H₁₂N₂ MW: 160.22 g/mol |
MDL Number:
MFCD11556357 CAS Number: 709649-72-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...