N-[(2-Chloro-1H-indol-3-yl)methyl]-N-methylamine ≥97%

Supplier: Matrix Scientific

081940-1G
102836-160EA 1118.46 USD
102836-160
N-[(2-Chloro-1H-indol-3-yl)methyl]-N-methylamine ≥97%
N-[(2-Chloro-1H-indol-3-yl)methyl]-N-methylamine

Hazard: Irritant

Formula: C₁₀H₁₁ClN₂
MDL Number: MFCD11505010
CAS Number: 1216775-82-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR