N-[(5-Chloro-2-phenyl-1H-imidazol-4-yl)methyl]-N-methylamine ≥95%
Supplier: Matrix Scientific
081931-1G
102836-142EA
920
USD
102836-142
N-[(5-Chloro-2-phenyl-1H-imidazol-4-yl)methyl]-N-methylamine ≥95%
N-[(5-Chloro-2-phenyl-1H-imidazol-4-yl)methyl]-N-methylamine
Hazard: Irritant
Formula:
C₁₁H₁₂ClN₃ |
MDL Number:
MFCD16622237 CAS Number: 1287218-53-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...