N-Methyl-N-[(2-methyl-1H-imidazol-4-yl)methyl]amine ≥97%
Supplier: Matrix Scientific
081930-1G
102836-140EA
1235.38
USD
102836-140
N-Methyl-N-[(2-methyl-1H-imidazol-4-yl)methyl]amine ≥97%
N-Methyl-N-[(2-methyl-1H-imidazol-4-yl)methyl]amine
Hazard: Irritant
Formula:
C₆H₁₁N₃ |
MDL Number:
MFCD14666466 CAS Number: 1287217-50-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...