N-Methyl-N-[(5-nitro-2-furyl)methyl]amine ≥97%

Supplier: Matrix Scientific

081911-1G
102836-100EA 949.23 USD
102836-100
N-Methyl-N-[(5-nitro-2-furyl)methyl]amine ≥97%
N-Methyl-N-[(5-nitro-2-furyl)methyl]amine

Hazard: Irritant

Formula: C₆H₈N₂O₃
MDL Number: MFCD12048862
CAS Number: 806604-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR