N-[(2,6-Difluoropyridin-3-yl)methyl]-N-methylamine ≥97%

Supplier: Matrix Scientific

081895-1G
102836-068EA 920 USD
102836-068
N-[(2,6-Difluoropyridin-3-yl)methyl]-N-methylamine ≥97%
N-[(2,6-Difluoropyridin-3-yl)methyl]-N-methylamine

Hazard: Irritant

Formula: C₇H₈F₂N₂
MDL Number: MFCD18089189
CAS Number: 1287217-70-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR