[6-(1H-Imidazol-1-yl)pyridin-2-yl]methylamine dihydrochloride ≥95%
Supplier: Matrix Scientific
081879-1G
102836-030EA
707.69
USD
102836-030
[6-(1H-Imidazol-1-yl)pyridin-2-yl]methylamine dihydrochloride ≥95%
[6-(1H-Imidazol-1-yl)pyridin-2-yl]methylamine dihydrochloride
Hazard: Irritant
Formula:
C₉H₁₂Cl₂N₄ |
MDL Number:
MFCD08704497 CAS Number: 1215840-38-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...