[2-(Cyclohexyloxy)pyridin-4-yl]methylamine ≥95%

Supplier: Matrix Scientific

081875-1G
102836-022EA 566.15 USD
102836-022
[2-(Cyclohexyloxy)pyridin-4-yl]methylamine ≥95%
[2-(Cyclohexyloxy)pyridin-4-yl]methylamine

Hazard: Irritant

Formula: C₁₂H₁₈N₂O
MDL Number: MFCD09258825
CAS Number: 953735-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR