[6-(2-Aziridin-1-ylethoxy)pyridin-3-yl]methylamine ≥95%

Supplier: Matrix Scientific

081872-1G
102836-016EA 566.15 USD
102836-016
[6-(2-Aziridin-1-ylethoxy)pyridin-3-yl]methylamine ≥95%
[6-(2-Aziridin-1-ylethoxy)pyridin-3-yl]methylamine

Hazard: Irritant

Formula: C₁₀H₁₅N₃O
MDL Number: MFCD09258822
CAS Number: 1086379-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR