[2-(2-Aziridin-1-ylethoxy)pyridin-4-yl]methylamine ≥95%

Supplier: Matrix Scientific

081871-1G
102836-014EA 566.15 USD
102836-014
[2-(2-Aziridin-1-ylethoxy)pyridin-4-yl]methylamine ≥95%
[2-(2-Aziridin-1-ylethoxy)pyridin-4-yl]methylamine

Hazard: Irritant

Formula: C₁₀H₁₅N₃O
MDL Number: MFCD09258821
CAS Number: 1086379-34-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR