[2-(1H-Imidazol-1-yl)pyridin-4-yl]methylamine ≥95%
Supplier: Matrix Scientific
081869-1G
102836-010EA
566.15
USD
102836-010
[2-(1H-Imidazol-1-yl)pyridin-4-yl]methylamine ≥95%
[2-(1H-Imidazol-1-yl)pyridin-4-yl]methylamine
Hazard: Irritant
Formula:
C₉H₁₀N₄ |
MDL Number:
MFCD09258819 CAS Number: 953731-28-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...