(2-Piperazin-1-ylpyridin-4-yl)methylamine ≥95%

Supplier: Matrix Scientific

081868-1G
102836-008EA 1118.46 USD
102836-008
(2-Piperazin-1-ylpyridin-4-yl)methylamine ≥95%
(2-Piperazin-1-ylpyridin-4-yl)methylamine

Hazard: Irritant

Formula: C₁₀H₁₆N₄
MDL Number: MFCD09258818
CAS Number: 1086379-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR