tert-Butyl-5-(aminomethyl)-6-methylpyridin-2-ylcarbamate ≥95%

081867-1G
102836-006EA 787.15 USD
102836-006
tert-Butyl-5-(aminomethyl)-6-methylpyridin-2-ylcarbamate ≥95%
tert-Butyl-5-(aminomethyl)-6-methylpyridin-2-ylcarbamate

Hazard: Irritant

Formula: C₁₂H₁₉N₃O₂
MW: 237.3 g/mol
MDL Number: MFCD09258817
CAS Number: 187163-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR