N-Methyl-N-[2-(4-phenoxyphenyl)ethyl]amine ≥97%

Supplier: Matrix Scientific

081856-1G
102835-980EA 949.23 USD
102835-980
N-Methyl-N-[2-(4-phenoxyphenyl)ethyl]amine ≥97%
N-Methyl-N-[2-(4-phenoxyphenyl)ethyl]amine

Hazard: Irritant

Formula: C₁₅H₁₇NO
MDL Number: MFCD12827854
CAS Number: 1219155-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR