N-[2-(4-Isopropylphenyl)ethyl]-N-methylamine ≥97%

Supplier: Matrix Scientific

081854-1G
102835-976EA 949.23 USD
102835-976
N-[2-(4-Isopropylphenyl)ethyl]-N-methylamine ≥97%
N-[2-(4-Isopropylphenyl)ethyl]-N-methylamine

Hazard: Irritant

Formula: C₁₂H₁₉N
MDL Number: MFCD12827852
CAS Number: 137069-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR