[1-(2,6-Difluorophenyl)cyclopropyl]methylamine ≥95%

Supplier: Matrix Scientific

081835-1G
102835-936EA 636.92 USD
102835-936
[1-(2,6-Difluorophenyl)cyclopropyl]methylamine ≥95%
[1-(2,6-Difluorophenyl)cyclopropyl]methylamine

Hazard: Irritant

Formula: C₁₀H₁₁F₂N
MDL Number: MFCD11100030
CAS Number: 1216908-25-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR