N-Methyl-N-(4-{[2-(trifluoromethyl)pyrimidin-4-yl]oxy}benzyl)amine ≥97%

Supplier: Matrix Scientific

081821-1G
102835-904EA 723.08 USD
102835-904
N-Methyl-N-(4-{[2-(trifluoromethyl)pyrimidin-4-yl]oxy}benzyl)amine ≥97%
N-Methyl-N-(4-{[2-(trifluoromethyl)pyrimidin-4-yl]oxy}benzyl)amine

Hazard: Irritant

Formula: C₁₃H₁₂F₃N₃O
MW: 283.26 g/mol
MDL Number: MFCD09800576
CAS Number: 1086379-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR