N-(2-Fluoro-4-methylbenzyl)-N-methylamine ≥97%

Supplier: Matrix Scientific

081786-1G
102835-814EA 863.08 USD
102835-814
N-(2-Fluoro-4-methylbenzyl)-N-methylamine ≥97%
N-(2-Fluoro-4-methylbenzyl)-N-methylamine

Hazard: Irritant

Formula: C₉H₁₂FN
MDL Number: MFCD12824256
CAS Number: 1178323-61-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR