N-Methyl-N-[3-(pentafluorosulfanyl)benzyl]amine ≥95%

Supplier: Matrix Scientific

081785-1G
102835-812EA 1118.46 USD
102835-812
N-Methyl-N-[3-(pentafluorosulfanyl)benzyl]amine ≥95%
N-Methyl-N-[3-(pentafluorosulfanyl)benzyl]amine

Hazard: Irritant

Formula: C₈H₁₀F₅NS
MDL Number: MFCD18089188
CAS Number: 1287218-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR