N-(2-Fluoro-3-methoxybenzyl)-N-methylamine ≥97%

Supplier: Matrix Scientific

081780-1G
102835-802EA 863.08 USD
102835-802
N-(2-Fluoro-3-methoxybenzyl)-N-methylamine ≥97%
N-(2-Fluoro-3-methoxybenzyl)-N-methylamine

Hazard: Irritant

Formula: C₉H₁₂FNO
MDL Number: MFCD18089183
CAS Number: 921219-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR