N-[2,5-Bis(trifluoromethyl)benzyl]-N-methylamine ≥97%
Supplier: Matrix Scientific
081779-1G
102835-800EA
863.08
USD
102835-800
N-[2,5-Bis(trifluoromethyl)benzyl]-N-methylamine ≥97%
N-[2,5-Bis(trifluoromethyl)benzyl]-N-methylamine
Hazard: Irritant
Formula:
C₁₀H₉F₆N |
MDL Number:
MFCD18089182 CAS Number: 958863-59-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...