N-[2,4-Bis(trifluoromethyl)benzyl]-N-methylamine ≥97%
Supplier: Matrix Scientific
081778-1G
102835-798EA
863.08
USD
102835-798
N-[2,4-Bis(trifluoromethyl)benzyl]-N-methylamine ≥97%
N-[2,4-Bis(trifluoromethyl)benzyl]-N-methylamine
Hazard: Irritant
Formula:
C₁₀H₉F₆N |
MDL Number:
MFCD13249471 CAS Number: 958863-74-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...