2-[(Methylamino)methyl]benzene-1,4-diol ≥97%

Supplier: Matrix Scientific

081768-1G
102835-776EA 553.85 USD
102835-776
2-[(Methylamino)methyl]benzene-1,4-diol ≥97%
2-[(Methylamino)methyl]benzene-1,4-diol

Hazard: Irritant

Formula: C₈H₁₁NO₂
MDL Number: MFCD16622232
CAS Number: 229486-97-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR