tert-Butyl (1R)-2-methyl-1-(2-oxoethyl)propylcarbamate ≥97%

Supplier: Matrix Scientific

081759-1G
102835-754EA 1372.31 USD
102835-754
tert-Butyl (1R)-2-methyl-1-(2-oxoethyl)propylcarbamate ≥97%
tert-Butyl (1R)-2-methyl-1-(2-oxoethyl)propylcarbamate

Hazard: Irritant

Formula: C₁₁H₂₁NO₃
MDL Number: MFCD21333117
CAS Number: 124754-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR