tert-Butyl-3-benzyl-3-formylpiperidine-1-carboxylate ≥95%
Supplier: Matrix Scientific
081730-1G
102835-668EA
1513.85
USD
102835-668
tert-Butyl-3-benzyl-3-formylpiperidine-1-carboxylate ≥95%
tert-Butyl-3-benzyl-3-formylpiperidine-1-carboxylate
Hazard: Irritant
Formula:
C₁₈H₂₅NO₃ MW: 303.4 g/mol |
MDL Number:
MFCD18064987 CAS Number: 1355334-49-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...