5-(1H-Imidazol-1-yl)-2-furaldehyde ≥97%

Supplier: Matrix Scientific

081663-1G
102835-504EA 566.15 USD
102835-504
5-(1H-Imidazol-1-yl)-2-furaldehyde ≥97%
5-(1H-Imidazol-1-yl)-2-furaldehyde

Hazard: Irritant

Formula: C₈H₆N₂O₂
MDL Number: MFCD09864906
CAS Number: 133531-41-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR