2-(4-Methylphenyl)isonicotinaldehyde ≥97%

Supplier: Matrix Scientific

081641-1G
102835-452EA 750.77 USD
102835-452
2-(4-Methylphenyl)isonicotinaldehyde ≥97%
2-(4-Methylphenyl)isonicotinaldehyde

Hazard: Irritant

Formula: C₁₃H₁₁NO
MDL Number: MFCD14666460
CAS Number: 1226166-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR