6-(4-Methylphenyl)pyridine-2-carbaldehyde ≥97%
Supplier: Matrix Scientific
081639-1G
102835-448EA
750.77
USD
102835-448
6-(4-Methylphenyl)pyridine-2-carbaldehyde ≥97%
6-(4-Methylphenyl)pyridine-2-carbaldehyde
Hazard: Irritant
Formula:
C₁₃H₁₁NO |
MDL Number:
MFCD06410022 CAS Number: 887979-33-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...