4-[(4-Trifluoromethyl)phenyl]pyridine-2-carbaldehyde ≥97%
Supplier: Matrix Scientific
081638-1G
102835-446EA
750.77
USD
102835-446
4-[(4-Trifluoromethyl)phenyl]pyridine-2-carbaldehyde ≥97%
4-[(4-Trifluoromethyl)phenyl]pyridine-2-carbaldehyde
Hazard: Irritant
Formula:
C₁₃H₈F₃NO MW: 251.21 g/mol |
MDL Number:
MFCD14666459 CAS Number: 1005189-47-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...