4-{[3-(Trifluoromethyl)pyridin-2-yl]oxy}benzaldehyde ≥97%

Supplier: Matrix Scientific

081616-1G
102835-400EA 496.92 USD
102835-400
4-{[3-(Trifluoromethyl)pyridin-2-yl]oxy}benzaldehyde ≥97%
4-{[3-(Trifluoromethyl)pyridin-2-yl]oxy}benzaldehyde

Hazard: Irritant

Formula: C₁₃H₈F₃NO₂
MDL Number: MFCD10703516
CAS Number: 1086379-05-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR