4-[(2-Chloropyrimidin-4-yl)oxy]benzaldehyde ≥95%
Supplier: Matrix Scientific
081597-1G
102835-362EA
806.15
USD
102835-362
4-[(2-Chloropyrimidin-4-yl)oxy]benzaldehyde ≥95%
4-[(2-Chloropyrimidin-4-yl)oxy]benzaldehyde
Hazard: Irritant
Formula:
C₁₁H₇ClN₂O₂ |
MDL Number:
MFCD09864903 CAS Number: 1086378-89-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...