2-Methoxy-5-pyrimidin-5-ylbenzaldehyde ≥97%

Supplier: Matrix Scientific

081594-1G
102835-352EA 486.15 USD
102835-352
2-Methoxy-5-pyrimidin-5-ylbenzaldehyde ≥97%
2-Methoxy-5-pyrimidin-5-ylbenzaldehyde

Hazard: Irritant

Formula: C₁₂H₁₀N₂O₂
MDL Number: MFCD09258811
CAS Number: 190271-69-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR