4-(Tetrahydro-2H-pyran-4-ylmethoxy)benzaldehyde ≥97%

Supplier: Matrix Scientific

081578-1G
102835-312EA 1153.85 USD
102835-312
4-(Tetrahydro-2H-pyran-4-ylmethoxy)benzaldehyde ≥97%
4-(Tetrahydro-2H-pyran-4-ylmethoxy)benzaldehyde

Hazard: Irritant

Formula: C₁₃H₁₆O₃
MDL Number: MFCD08669633
CAS Number: 1086378-74-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR