3,5-Difluoro-4-(trifluoromethyl)benzaldehyde ≥97%
Supplier: Matrix Scientific
081574-1G
102835-300EA
1429.23
USD
102835-300
3,5-Difluoro-4-(trifluoromethyl)benzaldehyde ≥97%
3,5-Difluoro-4-(trifluoromethyl)benzaldehyde
Hazard: Irritant
Formula:
C₈H₃F₅O |
MDL Number:
MFCD22566356 CAS Number: 1417569-98-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...