4-(3,5-Dimethylisoxazol-4-yl)benzaldehyde ≥95%
Supplier: Matrix Scientific
081540-1G
102835-218EA
750.77
USD
102835-218
4-(3,5-Dimethylisoxazol-4-yl)benzaldehyde ≥95%
4-(3,5-Dimethylisoxazol-4-yl)benzaldehyde
Hazard: Irritant
Formula:
C₁₂H₁₁NO₂ |
MDL Number:
MFCD12026346 CAS Number: 630392-25-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...