3-{[4-(Trifluoromethyl)pyrimidin-2-yl]oxy}benzaldehyde ≥95%

Supplier: Matrix Scientific

081537-1G
102835-210EA 723.08 USD
102835-210
3-{[4-(Trifluoromethyl)pyrimidin-2-yl]oxy}benzaldehyde ≥95%
3-{[4-(Trifluoromethyl)pyrimidin-2-yl]oxy}benzaldehyde

Hazard: Irritant

Formula: C₁₂H₇F₃N₂O₂
MDL Number: MFCD11505008
CAS Number: 1216892-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR