tert-Butyl-4-[2-(4-formylphenoxy)ethyl]piperazine-1-carboxylate ≥97%

Supplier: Matrix Scientific

081529-1G
102835-186EA 750.77 USD
102835-186
tert-Butyl-4-[2-(4-formylphenoxy)ethyl]piperazine-1-carboxylate ≥97%
tert-Butyl-4-[2-(4-formylphenoxy)ethyl]piperazine-1-carboxylate

Hazard: Irritant

Formula: C₁₈H₂₆N₂O₄
MW: 334.42 g/mol
MDL Number: MFCD11042248
CAS Number: 1086378-39-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR