3-Amino-6-chloro-1-methyl-1H-indazole ≥97%

Supplier: Matrix Scientific

081475-1G
102835-024EA 806.15 USD
102835-024
3-Amino-6-chloro-1-methyl-1H-indazole ≥97%
3-Amino-6-chloro-1-methyl-1H-indazole

Hazard: Irritant

Formula: C₈H₈ClN₃
MDL Number: MFCD09991761
CAS Number: 1031927-22-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR