tert-Butyl-2,3-dihydro-1H-inden-5-ylcarbamate ≥97%
Supplier: Matrix Scientific
081471-1G
102835-016EA
327.69
USD
102835-016
tert-Butyl-2,3-dihydro-1H-inden-5-ylcarbamate ≥97%
tert-Butyl-2,3-dihydro-1H-inden-5-ylcarbamate
Hazard: Irritant
Formula:
C₁₄H₁₉NO₂ MW: 233.31 g/mol |
MDL Number:
MFCD09991759 CAS Number: 695231-56-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...