tert-Butyl-3-chloropropyl(cyclopropyl)carbamate ≥97%
Supplier: Matrix Scientific
081442-1G
102834-948EA
723.08
USD
102834-948
tert-Butyl-3-chloropropyl(cyclopropyl)carbamate ≥97%
tert-Butyl-3-chloropropyl(cyclopropyl)carbamate
Hazard: Irritant
Formula:
C₁₁H₂₀ClNO₂ MW: 233.74 g/mol |
MDL Number:
MFCD09800569 CAS Number: 1000018-27-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...