N-(2,4-Dinitrophenyl)-1-methylpiperidin-4-amine ≥97%

Supplier: Matrix Scientific

081407-1G
102834-826EA 949.23 USD
102834-826
N-(2,4-Dinitrophenyl)-1-methylpiperidin-4-amine ≥97%
N-(2,4-Dinitrophenyl)-1-methylpiperidin-4-amine

Hazard: Irritant

Formula: C₁₂H₁₆N₄O₄
MDL Number: MFCD22126063
CAS Number: 925216-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR