5-Pyrazin-2-yl-1,2,4-oxadiazol-3-amine ≥97%

Supplier: Matrix Scientific

081378-1G
102834-748EA 723.08 USD
102834-748
5-Pyrazin-2-yl-1,2,4-oxadiazol-3-amine ≥97%
5-Pyrazin-2-yl-1,2,4-oxadiazol-3-amine

Hazard: Irritant

Formula: C₆H₅N₅O
MDL Number: MFCD09864899
CAS Number: 1086378-64-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR